missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10341563
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10341563

Brand: Thermo Scientific Maybridge CC31009DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 864068-79-1
Molecular Formula C14H22N2O3S
Molecular Weight (g/mol) 298.401
InChI Key RDZWXODJIWMJPE-UHFFFAOYSA-N
Synonym tert-butyl 4-4-hydroxymethyl-1,3-thiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-hydroxymethyl thiazol-2-yl piperidine-1-carboxylate, 4-4-hydroxymethyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 1-1,1-dimethylethyl 4-4-hydroxymethyl-2-thiazolyl-1-piperidine-carboxylate, 1-1,1-dimethylethyl 4-4-hydroxymethyl-2-thiazolyl-1-piperidinecarboxylate, 1-piperidinecarboxylicacid, 4-4-hydroxymethyl-2-thiazolyl-, 1,1-dimethylethyl ester
PubChem CID 7164573
IUPAC Name tert-butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C2=NC(=CS2)CO

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 298.4
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-[4-(hydroxymethyl)-1, 3-thiazol-2-yl]piperidine-1-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.