missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10285714
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10285714 5 g 5g
10505963 250 mg 250mg
10341563 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10285714 Supplier Thermo Scientific Maybridge Supplier No. CC31009DE
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 864068-79-1
Molecular Formula C14H22N2O3S
Molecular Weight (g/mol) 298.401
InChI Key RDZWXODJIWMJPE-UHFFFAOYSA-N
Synonym tert-butyl 4-4-hydroxymethyl-1,3-thiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-hydroxymethyl thiazol-2-yl piperidine-1-carboxylate, 4-4-hydroxymethyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 1-1,1-dimethylethyl 4-4-hydroxymethyl-2-thiazolyl-1-piperidine-carboxylate, 1-1,1-dimethylethyl 4-4-hydroxymethyl-2-thiazolyl-1-piperidinecarboxylate, 1-piperidinecarboxylicacid, 4-4-hydroxymethyl-2-thiazolyl-, 1,1-dimethylethyl ester
PubChem CID 7164573
IUPAC Name tert-butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C2=NC(=CS2)CO

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 298.4
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-[4-(hydroxymethyl)-1, 3-thiazol-2-yl]piperidine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.