missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyraz, 97%, Maybridge

Product Code. 10708065
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10708065 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10708065 Supplier Thermo Scientific Maybridge Supplier No. MAY00063DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1073354-70-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C16H27BN2O4
MDL Number MFCD09027070
Quantity 1 g
Synonym tert-butyl 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-carboxylate, 1-boc-3,5-dimethylpyrazole-4-boronic acid pinacol ester, tert-butyl 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole-1-carboxylate, 1-tert-butoxycarbonyl-3,5-dimethylpyrazole-4-boronic acid, pinacol ester, 1-tert-butoxycarbonyl-3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 3,5-dimethyl-1h-pyrazole-4-boronic acid, pinacol ester, n1-boc protected, 1-tert-butoxycarbonyl-3,5-dimethyl-1h-pyrazol-4-yl boronic acid pinacol ester, tert-butyl 3,5-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole-1-carboxylate, 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrazole-1-carboxylic acid tert-butyl ester
InChI Key GNLNGVZPEKSDAR-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N(N=C2C)C(=O)OC(C)(C)C)C
IUPAC Name tert-butyl 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylate
Molecular Weight (g/mol) 322.212
PubChem CID 16218387
Formula Weight 322.21
Percent Purity 97%
Chemical Name or Material tert-Butyl 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.