missing translation for 'onlineSavingsMsg'
Learn More

tert-Butoxybis(dimethylamino)methane

Product Code. p-3213175 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10145272

Brand: Thermo Scientific Acros 378120050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5815-08-7 | C9H22N2O | 174.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5815-08-7
Molecular Formula C9H22N2O
Molecular Weight (g/mol) 174.29
MDL Number MFCD00042858
InChI Key HXRAMSFGUAOAJR-UHFFFAOYSA-P
Synonym tert-butoxy bis dimethylamino methane, 1-tert-butoxy-n,n,n',n'-tetramethylmethanediamine, tert-butoxybis dimethylamino methane, bredereck's reagent, bredereck reagent, tert-butoxy dimethylamino methyl dimethylamine, unii-jej0u558f7, bis dimethylamino-t-butoxymethane, tert-butoxy-bis dimethylamino methane, bis dimethylamino tert-butoxy methane
PubChem CID 79885
IUPAC Name N,N,N',N'-tetramethyl-1-[(2-methylpropan-2-yl)oxy]methanediamine
SMILES CC(C)(C)OC(N(C)C)N(C)C

Specifications

Color Yellow
Density 0.8440g/mL
Boiling Point 50°C to 55°C (15.0 mmHg)
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 85+%
Packaging Glass bottle
Linear Formula (CH3)3COCH[N(CH3)2]2
Refractive Index 1.4220 to 1.425
Quantity 5 g
Merck Index 15, 1373
Specific Gravity 0.844
Solubility Information Solubility in water: decomposes
Formula Weight 174.29
Percent Purity 90% (sum of 3 constituents) varying proportionally: Tris(dimethylamino)methane, N,N-dimethylformamide-N,N-di-tert.-butylacetal, tert.-Butoxybis(dimethylamino)methane
Physical Form Liquid
Chemical Name or Material tert-Butoxybis(dimethylamino)methane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butoxybis(dimethylamino)methane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.