Learn More
Tentoxin, TRC
Specifications
Specifications
| Chemical Name or Material | Tentoxin |
| CAS | 28540-82-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H30 N4 O4 |
| Quantity | 2.5 mg |
| Synonym | Cyclo(N-methyl-L-alanyl-L-leucyl-α,β-didehydro-N-methylphenylalanylglycyl)','(Z)-, Tentoxin (8CI), 1,4,7,10-Tetraazacyclododecane','cyclic peptide deriv., Cyclic(N-methyl-L-alanyl-L-leucyl-α,β-didehydro-N-methylphenylalanylglycyl)','(Z)- |
| InChI Formula | InChI=1S/C22H30N4O4/c1-14(2)11-17-22(30)26(5)18(12-16-9-7-6-8-10-16)21(29)23-13-19(27)25(4)15(3)20(28)24-17/h6-10,12,14-15,17H,11,13H2,1-5H3,(H,23,29)(H,24,28)/b18-12-/t15-,17-/m0/s1 |
| SMILES | CC(C)C[C@@H]1NC(=O)[C@H](C)N(C)C(=O)CNC(=O)\C(=C\c2ccccc2)\N(C)C1=O |
| IUPAC Name | (3S,6S,12Z)-12-benzylidene-1,6,7-trimethyl-3-(2-methylpropyl)-1,4,7,10-tetrazacyclododecane-2,5,8,11-tetrone |
| Molecular Weight (g/mol) | 414.5 |
| Show More |