missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tensin 4 Monoclonal Antibody (SP83)
GREENER_CHOICE

Product Code. 13441087
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13441087 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13441087 Supplier Invitrogen™ Supplier No. MA516355

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is liver hepatocellular carcinoma.

TNS4 links the focal adhesion and cell adhesion signaling to the actin cytoskeleton. It may be involved in cell survival and migration. The protein may be involved in cell migration, cartilage development and in linking signal transduction pathways to the cytoskeleton and promote apoptosis, via its cleavage by caspase-3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tensin 4
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP83
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene TNS4
Gene Accession No. Q8IZW8
Gene Alias 9930017A07Rik; AA589547; AU016405; C terminal tensin like; CTEN; c-ten; C-terminal tensin-like protein; PP14434; RGD1310402; tensin 4; tensin-4; TNS4
Gene Symbols TNS4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of human CTEN protein.
Purification Method Protein A/G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84951
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.