Learn More
Tenovin-6, MedChemExpress
Product Code. 16419157
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Product Code. 16419157
Brand: Medchem Express HY15510
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Tenovin-6, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 also inhibits dihydroorotate dehydrogenase (DHODH).
Tenovin-6, an analog of Tenovin-1 (HY-13423), is an activator of p53 transcriptional activity. Tenovin-6 inhibits the protein deacetylase activities of purified human SIRT1, SIRT2, and SIRT3 with IC50s of 21 μM, 10 μM, and 67 μM, respectively. Tenovin-6 also inhibits dihydroorotate dehydrogenase (DHODH).
Specifications
Chemical Name or Material Tenovin-6
Physical Form Solid
CAS 1011557-82-6
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H34N4O2S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 31 mg/mL (68.19 mM)
SMILES O=C(NC(NC1=CC=C(NC(CCCCN(C)C)=O)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2
Molecular Weight (g/mol) 454.63
Formula Weight 454.63
Percent Purity 98.67%
Grade Research