Learn More
Tenofovir Isopropyl Carbamate, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Tenofovir Isopropyl Carbamate |
| CAS | 1391053-20-5 |
| Molecular Formula | C13H20N5O6P |
| Quantity | 10 mg |
| Synonym | [[(1R)-2(6-Isopropylaminocarbamate-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic Acid |
| InChI Formula | InChI=1S/C13H20N5O6P/c1-8(2)24-13(19)17-11-10-12(15-5-14-11)18(6-16-10)4-9(3)23-7-25(20,21)22/h5-6,8-9H,4,7H2,1-3H3,(H2,20,21,22)(H,14,15,17,19)/t9-/m1/s1 |
| SMILES | CC(C)OC(=O)Nc1ncnc2c1ncn2C[C@@H](C)OCP(=O)(O)O |
| IUPAC Name | [(2R)-1-[6-(propan-2-yloxycarbonylamino)purin-9-yl]propan-2-yl]oxymethylphosphonic acid |
| Molecular Weight (g/mol) | 373.3 |
| Formula Weight | 373.1151 |