Learn More
Tenofovir alafenamide hemifumarate, MedChemExpress
Product Code. 16498967
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16498967
Brand: Medchem Express HY15232B
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Tenofovir alafenamide hemifumarate (GS-7340 hemifumarate) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor.
Tenofovir alafenamide hemifumarate (GS-7340 hemifumarate) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor.
Specifications
Chemical Name or Material Tenofovir alafenamide hemifumarate
CAS 1392275-56-7
Color White
Purity Grade Notes Research
Molecular Formula C21H29N6O5P.1/2 C4H4O4
Quantity 5 mg
Synonym GS-7340 hemifumarate
Solubility Information DMSO : 50 mg/mL (93.55 mM; Need ultrasonic) ∣H2O : 20 mg/mL (37.42 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)/C=C/C(O)=O.NC1=NC=NC2=C1N=CN2C[C@@H](C)OC[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C)C)=O)=O.[0.5]
Molecular Weight (g/mol) 534.5
Formula Weight 534.5
Percent Purity 99.29%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus