Learn More
Tenidap, TRC
Specifications
Specifications
| Chemical Name or Material | Tenidap |
| CAS | 120210-48-2 |
| Molecular Formula | C14 H9 Cl N2 O3 S |
| Quantity | 25 mg |
| Synonym | 1H-Indole-1-carboxamide','5-chloro-2,3-dihydro-3-(hydroxy-2-thienylmethylene)-2-oxo-','(3Z)-, 1H-Indole-1-carboxamide','5-chloro-2,3-dihydro-3-(hydroxy-2-thienylmethylene)-2-oxo-','(Z)-, (3Z)-5-Chloro-2,3-dihydro-3-(hydroxy-2-thienylmethylene)-2-oxo-1H-indole-1-carboxamide, CP 66248, Tenidap |
| InChI Formula | InChI=1S/C14H9ClN2O3S/c15-7-3-4-9-8(6-7)11(13(19)17(9)14(16)20)12(18)10-2-1-5-21-10/h1-6,18H,(H2,16,20)/b12-11- |
| SMILES | NC(=O)N1C(=O)\C(=C(/O)\c2cccs2)\c3cc(Cl)ccc13 |
| IUPAC Name | (3Z)-5-chloro-3-[hydroxy(thiophen-2-yl)methylidene]-2-oxoindole-1-carboxamide |
| Molecular Weight (g/mol) | 320.75 |
| Formula Weight | 320.0022 |