missing translation for 'onlineSavingsMsg'
Learn More

Temuterkib, MedChemExpress

Product Code. 16450769
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16450769

Brand: Medchem Express HY101494

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Temuterkib (LY3214996) is a highly selective inhibitor of ERK1 and ERK2, with IC50 of 5 nM for both enzymes in biochemical assays. Temuterkib potently inhibits cellular p-RSK1 in BRAF and RAS mutant cancer cell lines. Temuterkib shows potent antitumor activities in cancer models with MAPK pathway alterations.

Temuterkib (LY3214996) is a highly selective inhibitor of ERK1 and ERK2, with IC50 of 5 nM for both enzymes in biochemical assays. Temuterkib potently inhibits cellular p-RSK1 in BRAF and RAS mutant cancer cell lines. Temuterkib shows potent antitumor activities in cancer models with MAPK pathway alterations.

Specifications

Chemical Name or Material Temuterkib
Physical Form Solid
CAS 1951483-29-6
Color Light Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C22H27N7O2S
For Use With (Application) Cancer-Kinase/protease
Synonym LY3214996
Solubility Information DMSO : 20 mg/mL (44.10 mM; Need ultrasonic)
SMILES O=C(C(C=C(C1=NC(NC2=CC=NN2C)=NC=C1)S3)=C3C4(C)C)N4CCN5CCOCC5
Molecular Weight (g/mol) 453.56
Formula Weight 453.56
Percent Purity 99.54%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.