Learn More
Eppendorf™ Temperature Verification System, USB, Single Channel
Shop All Eppendorf Products
Click to view available options
:
For use with Mastercycler
Description
- Measuring device without temperature sensor
- Sensor for 96 well and 384 well blocks, sold separately
- Simple, rapid verification and adjustment
- Automatic adjustment with Mastercycler software, version 2.10 or higher
- Can be third-party adjusted
Specifications
Specifications
| Description | USB, single-channel |
| Product Type | Temperature Verification System |