Learn More
Edmund Bühler GmbH™ Temperature Range Extender
Product Code. 10145651
Click to view available options
For Use With (Equipment):
Incubator Hoods TH 15 and TH 30
Unit Size:
Each
Product Code. 10145651
Brand: Edmund Bühler GmbH™ 0052 105
Only null left
Add to basket
Add to basket
Extend temperature range up to 60°C with an additional heater. Edmund Bühler GmbH™ Temperature Range Extender is for use with an incubator hood.
  • Additional heater for incubator hood TH 15 and TH 30
  • Extends temperature up to 60°C
  • Installation only possible in Edmund Bühler GmbH™ works
Specifications
Temperature Range (Metric) 60°C (Max.)
For Use With (Equipment) Incubator Hoods TH 15 and TH 30