Learn More
Edmund Bühler GmbH™ Temperature Range Extender
Click to view available options
For Use With (Equipment):
Incubator Hoods TH 15 and TH 30
Unit Size:
Each
Description
- Additional heater for incubator hood TH 15 and TH 30
- Extends temperature up to 60°C
- Installation only possible in Edmund Bühler GmbH™ works
Specifications
Specifications
| Temperature Range (Metric) | 60°C (Max.) |
| For Use With (Equipment) | Incubator Hoods TH 15 and TH 30 |