Learn More
Dostmann Electronic™ Immersion Temperature Probe
Click to view available options
Length (Metric):
130 mm
220 mm
300 mm
Temperature Range (Metric):
-100°C to +1100°C
-200°C to +1100°C
Diameter (Metric) Probe:
1 mm
1.5 mm
3 mm
Unit Size:
Each
Description
- Type K according to EN 60584-1
- NiCr-Ni
- Mineral-insulated
Specifications
Specifications
| Diameter (Metric) Probe | 3 mm |
| For Use With (Application) | For Measuring In Liquid And Powdered Materials |
| Includes | No handle, or cable |
| Length (Metric) | 220 mm |
| Product Type | Temperature Probe |
| Description | Flexible probe |
| Temperature Range (Metric) | -200°C to +1100°C |
| For Use With (Equipment) | P410 / TC301 / TC 309 / Mini-K |