missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Telratolimod, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
Telratolimod (MEDI9197) is a potent toll like receptors 7/8 (TLR7/8) agonist, with antitumor activity.
Specifications
Specifications
| Chemical Name or Material | Telratolimod |
| CAS | 1359993-59-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H59N5O2 |
| Quantity | 10 mg |
| Synonym | MEDI9197 3M-052 |
| Solubility Information | DMSO : 62.5 mg/mL (105.24 mM; Need ultrasonic) |
| SMILES | NC1=NC2=CC=CC=C2C3=C1N=C(CCCC)N3OCCCCNC(CCCCCCCCCCCCCCCCC)=O |
| Molecular Weight (g/mol) | 593.89 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction