Learn More
Telotristat, MedChemExpress
Product Code. 16410196
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
Product Code. 16410196
Brand: Medchem Express HY13055B
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Telotristat (LP-778902) is a potent tryptophan hydroxylase inhibitor with an in vivo IC50 of 0.028 μM.
Telotristat (LP-778902) is a potent tryptophan hydroxylase inhibitor with an in vivo IC50 of 0.028 μM.
Specifications
Chemical Name or Material Telotristat
CAS 1033805-28-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H22ClF3N6O3
Quantity 2 mg
Synonym LP-778902
Solubility Information DMSO : 33.33 mg/mL (60.94 mM; Need ultrasonic)
SMILES N[C@@H](CC1=CC=C(C2=NC(N)=NC(O[C@H](C3=CC=C(Cl)C=C3N4N=C(C)C=C4)C(F)(F)F)=C2)C=C1)C(O)=O
Molecular Weight (g/mol) 546.93
Formula Weight 546.93
Percent Purity 98.91%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation