missing translation for 'onlineSavingsMsg'
Learn More

Telotristat ethyl, MedChemExpress

Product Code. 16435518
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16435518

Brand: Medchem Express HY13055A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Telotristat ethyl (LX1032) is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production.

Telotristat ethyl (LX1032) is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production.

Specifications

Chemical Name or Material Telotristat ethyl
CAS 1033805-22-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H26ClF3N6O3
Quantity 10 mg
Synonym LX1032LX1606
Solubility Information DMSO : ≥ 101.5 mg/mL (176.53 mM)
SMILES N[C@@H](CC1=CC=C(C2=NC(N)=NC(O[C@H](C3=CC=C(Cl)C=C3N4N=C(C)C=C4)C(F)(F)F)=C2)C=C1)C(OCC)=O
Molecular Weight (g/mol) 574.98
Formula Weight 574.98
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.