missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) ethoxide, tech. 85%, remainder ethanol

Product Code. 11319527
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319527 2 g 2g
11309527 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319527 Supplier Thermo Scientific Alfa Aesar Supplier No. 041105.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2017-01-8 | C6H14N2O | 130.19 g/mol

Tellurium(IV) ethoxide could be used in sol-gel processing for the fabrication of tellurite thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2017-01-8
MDL Number MFCD00270937

Specifications

Melting Point >100°C (decomposition)
Density 1.52 g/mL
Boiling Point 88°C to 90°C (2 mmHg)
Flash Point 27°C (81°F)
Odor Alcohol-like
Quantity 2 g
UN Number UN1993
Sensitivity Light and moisture sensitive
Synonym tetraethoxytellurium iv, tellurium iv ethoxide, triethoxy-??-tellanyl oxy ethane
Solubility Information Decomposes in water to TeO2
Formula Weight 307.84
Percent Purity 85%
Grade Technical
Physical Form Liquid
Assay Remainder ethanol
Chemical Name or Material Tellurium(IV) ethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.