missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: MAQTVPPCEL PCKEYDVARN TGAYTSSGLA TASFRTSKYL LEEWFQNCYA RYHQAFADRD QSERQRHESQ QLATETQA Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.
TEKT4 is structural component of ciliary and flagellar microtubules. TEKT4 forms filamentous polymers in the walls of ciliary and flagellar microtubules.
Specifications
Specifications
| Antigen | TEKT4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TEKT4 |
| Gene Accession No. | Q8WW24 |
| Gene Alias | 1700010L19Rik; RGD1308075; Tek4; Tekt4; tektin 4; Tektin4; tektin-4; Testicular microtubules-related protein 4 |
| Gene Symbols | TEKT4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
