Learn More
behr Labor-Technik™ Technik™ Stand
Click to view available options
For Use With (Application):
12 Cod Air Coolers
Unit Size:
Each
Description
- Stand for 12 COD air condensers
- Damping elements made of rubber prevent glass from breaking when inserting it in the stand
Specifications
Specifications
| Product Type | Stand |
| Description | LS 12 Stand for 12 COD air condensers |
| For Use With (Application) | 12 Cod Air Coolers |
| Quantity | 1 Ea. |