Learn More
Technico™ Pocket Refractometer
Product Code. p-8009746
Click to view available options
Scale Range:
0 to 10 Brix
0 to 100 Salinity
0 to 15 Brix
28 to 62 Brix
Unit Size:
Each
Product Code. 10156362
Brand: Technico 8100.0150 M
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Pocket Refractometer
Scale Range 0 to 15 Brix
Refractive Index 1.333 to 1.356
Model DIGIT-015
Calibration Zero calibration with distilled water at 20°C
For Use With (Application) Measuring low sugar concentrations and low refractive values
Weight (Metric) 160 g
Dimensions (L x W x H) 55 x 80 x 205 mm