Learn More
Medline™ Technico™ DIGIT-025 ATC Wine Pocket Refractometer
Click to view available options
Unit Size:
Each
Specifications
Specifications
| Type | Wine Pocket Refractometer |
| Scale Range | 0 to 25 Brix |
| Model | DIGIT-025 ATC |
| Calibration | Zero calibration with distilled water between 10 and 30°C. |
| Quantity | 1 Each |