missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEAD1/2/3/4 Recombinant Rabbit Monoclonal Antibody (23GB4680), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
195.00 € - 446.00 €
Specifications
| Antigen | TEAD1/2/3/4 |
|---|---|
| Clone | 23GB4680 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088114
![]() |
Invitrogen™
MA552728 |
100 μL |
446.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089152
|
Invitrogen™
MA552729 |
20 μL |
195.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TEAD1/2/3/4 Recombinant Monoclonal antibody specifically detects TEAD1/2/3/4 in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| TEAD1/2/3/4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| P28347 | |
| 361630, 7003 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4680 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| DTEF-1; ETF; ETFR-1; Protein GT-IIC; RTEF1; RTEF-1; TCF13; TCF-13; TEA Domain Family Member 1; TEA Domain Family Member 1 (SV40 Transcriptional Enhancer Factor); TEA Domain Family Member 2; TEA Domain Family Member 3; TEA Domain Family Member 5; TEA Domain Transcription Factor 1; TEA Domain Transcription Factor 2; TEA Domain Transcription Factor 3; TEAD; TEAD2; TEAD-2; TEAD3; TEAD-3; TEAD-4; TEAD5; TEF3; TEF4; TEF-4; TEF5; TEF-5; Transcription Factor 13; Transcriptional Enhancer Factor 1; Transcriptional Enhancer Factor 1-Related; Transcriptional Enhancer Factor 5; Transcriptional Enhancer Factor TEF-1; Transcriptional Enhancer Factor TEF-4; Transcriptional Enhancer Factor TEF-5; Transcriptional Enhancer Factor TEF-5 (DTEF-1) | |
| A synthesized peptide derived from human TEAD1+TEAD2+TEAD3+TEAD4 (150-300AA). | |
| Primary | |
| -20°C | |
| TEAD1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title