missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF4 Antibody (TCF4/2252) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14192JF525
This item is not returnable.
View return policy
Description
TCF4 Monoclonal antibody specifically detects TCF4 in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| TCF4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4 | |
| Recombinant human TCF4 protein fragment (around aa 365-671) (exact sequence is proprietary) (Uniprot: P15884 ) | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 6925 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| TCF4/2252 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction