Learn More
TC13172, MedChemExpress
Product Code. 16489926
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16489926
Brand: Medchem Express HY101524
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress TC13172 is a mixed lineage kinase domain-like protein (MLKL) inhibitor with an EC50 value of 2 nM for HT-29 cells.
TC13172 is a mixed lineage kinase domain-like protein (MLKL) inhibitor with an EC50 value of 2 nM for HT-29 cells.
Specifications
Chemical Name or Material TC13172
CAS 2093393-05-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H16N4O5S
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (64.37 mM; Need ultrasonic)
SMILES O=C1C2=C(N=C(S(C)(=O)=O)N2C)N(CC#CC3=CC(O)=CC=C3)C(N1C)=O
Molecular Weight (g/mol) 388.4
Formula Weight 388.4
Percent Purity 98.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease