Learn More
Tocris Bioscience™ TC-S 7010
Description
TC-S 7010 is a potent and selective inhibitor of Aurora kinase A (IC50 = 3.4 nM); exhibits 1000-fold selectivity for Aurora kinase A over Aurora kinase B. Displays antiproliferative activity in HCT116 and HT29 cells (IC50 values are 0.19 and 2.9 μM respectively).
Specifications
Specifications
| Chemical Name or Material | N-(2-Chlorophenyl)-4-[[2-[[4-[2-(4-ethyl-1-piperazinyl)-2-oxoethyl]phenyl]amino]-5-fluoro-4-pyrimidinyl]amino]-benzamide |
| CAS | 1158838-45-9 |
| Quantity | 10 mg |
| Target | Aurora Kinase Inhibitors |
| Molecular Formula | C31H31ClFN7O2 |
| Formula Weight | 588.07 |
| Purity | >99% |