Learn More
Tocris Bioscience™ TC-S 7009
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
TC-S 7009 is a high affinity and selective HIF-2α inhibitor (Kd = 81 nM). Binds to the HIF-2α PAS-B domain; disrupts HIF-2α-ARNT heterodimerization. Exhibits >60-fold selectivity for HIF-2α over HIF-1α. Decreases HIF-2α DNA-binding and attenuates expression of HIF-2α target genes in vitro.
Specifications
Specifications
| Chemical Name or Material | N-(3-Chloro-5-fluorophenyl)-4-nitro-2,1,3-benzoxadiazol-5-amine |
| CAS | 1422955-31-4 |
| Assay Percent Range | >99% |
| Quantity | 5 mg |
| Molecular Formula | C12H6ClFN4O3 |
| Formula Weight | Observed MW: 308.65 |