missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-N 1752
GREENER_CHOICE

Product Code. 30001508
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001508 10 mg 10mg
18004242 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001508 Supplier Tocris Bioscience™ Supplier No. 4435/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NaV channel blocker

TC-N 1752 is a human NaV channel inhibitor (IC50 values are 0.17, 0.3, 0.4, 1.1 and 1.6 μM at hNaV1.7, hNaV1.3, hNaV1.4, hNaV1.5 and hNav1.9 respectively). Also inhibits tetrodotoxin -sensitive sodium channels. Displays analgesic efficacy in the formalin pain model.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1211866-85-1
Molecular Formula C25H27F3N6O3

Specifications

Quantity 10 mg
Formula Weight 516.52
Purity >99%
Molecular Weight (g/mol) 516.525
Chemical Name or Material TC-N 1752
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.