missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-DAPK 6

Product Code. 18014422
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18014422

Brand: Tocris Bioscience™ 4301/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective inhibitor of DAPK1

Potent and selective, ATP-competitive inhibitor of death-associated protein kinase 1 (DAPK1) (IC50 values are 69 and 225 nM for DAPK1 and DAPK3 respectively, when assayed with 10 μM ATP). Displays selectivity for DAPK1 over a range of 48 other kinases, including Abl, AMPK, Chk1, Met and Src (IC50 > 10 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 315694-89-4
Molecular Formula C17H12N2O2

Specifications

Quantity 50 mg
Formula Weight 276.29
Purity >98%
Molecular Weight (g/mol) 276.295
Chemical Name or Material TC-DAPK 6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.