Learn More
Tocris Bioscience™ TBK1 control PROTAC™ 4
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
TBK1 control PROTAC™ 4 is a negative control for TBK1 PROTAC™ 3i (Cat. No. 7259). Binds TBK1 with high affinity, but exhibits no significant degradation of TBK1. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2S,4S)-1-((S)-18-(4-((5-Bromo-4-((3-(N-methylcyclobutanecarboxamido)propyl)amino)pyrimidin-2-yl)amino)phenoxy)-2-(tert-butyl)-4-oxo-6,10,15-trioxa-3-azaoctadecanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2052306-31-5 |
| Quantity | 5 mg |
| Target | Active Degrader Negative Controls |
| Molecular Formula | C53H74BrN9O9S |
| Purity | 98% |