Learn More
Invitrogen™ TBCK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559007
Description
Immunogen sequence: LLANGFNECI LLFSDLPEID IERCVRESIN LFCWTPKSAT YRQHAQPPKP SSDSSGGRSS APYFSAECPD PPKTDLSRES IPLNDLKSEV SPRISAEDLI DLCELTVTGH FKTPS Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
Involved in the modulation of mTOR signaling and expression of mTOR complex components. Involved in the regulation of cell proliferation and growth. Involved in the control of actin-cytoskeleton organization.
Specifications
| TBCK | |
| Polyclonal | |
| Unconjugated | |
| TBCK | |
| 1700120J03Rik; 9430001M19; A630047E20Rik; C030007I09Rik; HSPC302; IHPRF3; RGD1307816; TBC domain-containing protein kinase-like protein; TBC1 domain containing kinase; TBCK; TBCKL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 93627 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TEA7 | |
| TBCK | |
| Recombinant protein corresponding to Human TBCK. Recombinant protein control fragment (Product #RP-98304). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.