missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAUT Polyclonal Antibody
GREENER_CHOICE

Product Code. 16613975
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16613975 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16613975 Supplier Invitrogen™ Supplier No. PA599725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAUT.

Taurine is an abundant organic osmolyte that possesses antioxidant and immunomodulatory properties and plays a role in cell volume homeostasis. Taurine is taken up into cells via the taurine transporter (TAUT). TAUT, which is sodium- and chloride-dependent, is a multi-pass membrane protein belonging to the sodium neurotransmitter symporter (SNF) family of proteins. TNF alpha upregulates TAUT expression, while phosphorylation on Serine 322 downregulates it. Overexpression of TAUT protects renal cells from cisplatin-induced nephrotoxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAUT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC6A6
Gene Accession No. P31641
Gene Alias AA589629; C80501; MGC10619; MGC131729; OTTHUMP00000208177; OTTHUMP00000208180; SC6A6; Slc6a6; Sodium- and chloride-dependent taurine transporter; Solute carrier 6 ,member 6 (taurine transporter); solute carrier family 6 (neurotransmitter transporter), member 6; solute carrier family 6 (neurotransmitter transporter, taurine), member 6; solute carrier family 6 member 6; solute carrier family 6, member 6; Taurine Transporter; taurine/beta-alanine transporter; TAUT
Gene Symbols SLC6A6
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC6A6(Accession P31641), corresponding to amino acid residues I563-H613.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6533
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.