missing translation for 'onlineSavingsMsg'
Learn More

Tauroursodeoxycholate sodium, MedChemExpress

Product Code. 16421906
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16421906

Brand: Medchem Express HY19696A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tauroursodeoxycholate (Tauroursodeoxycholic acid; TUDCA) sodium is an endoplasmic reticulum (ER) stress inhibitor. Tauroursodeoxycholate significantly reduces expression of apoptosis molecules, such as caspase-3 and caspase-12. Tauroursodeoxycholate also inhibits ERK.

Tauroursodeoxycholate (Tauroursodeoxycholic acid

Specifications

Chemical Name or Material Tauroursodeoxycholate sodium
CAS 35807-85-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H44NNaO6S
Quantity 500 mg
Synonym Tauroursodeoxycholic acid sodiumTUDCA sodiumUR 906 sodium
Solubility Information H2O : 100 mg/mL (191.68 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (191.68 mM; Need ultrasonic)
SMILES C[C@H](CCC(NCCS(=O)(O[Na])=O)=O)[C@H]1CC[C@@]2([H])[C@]3([H])[C@@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C
Molecular Weight (g/mol) 521.69
Formula Weight 521.69
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.