missing translation for 'onlineSavingsMsg'
Learn More

Taurodeoxycholic acid sodium hydrate, MedChemExpress

Product Code. p-200100147
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16444637

Brand: Medchem Express HYB1899A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Taurodeoxycholic acid sodium hydrate (Sodium taurodeoxycholate monohydrate) prevents apoptosis by blocking a calcium-mediated apoptotic pathway as well as caspase-12 activation. Taurodeoxycholic acid sodium hydrate is investigated for use in several conditions such as Primary Biliary Cirrhosis (PBC), insulin resistance, amyloidosis, Cystic Fibrosis, Cholestasis, and Amyotrophic Lateral Sclerosis.

Taurodeoxycholic acid sodium hydrate (Sodium taurodeoxycholate monohydrate) prevents apoptosis by blocking a calcium-mediated apoptotic pathway as well as caspase-12 activation. Taurodeoxycholic acid sodium hydrate is investigated for use in several conditions such as Primary Biliary Cirrhosis (PBC), insulin resistance, amyloidosis, Cystic Fibrosis, Cholestasis, and Amyotrophic Lateral Sclerosis.

Specifications

Chemical Name or Material Taurodeoxycholic acid sodium hydrate
CAS 110026-03-4
Color White
Purity Grade Notes Research
Molecular Formula C26H46NNaO7S
Quantity 50 mg
Synonym Sodium taurodeoxycholate monohydrate
Solubility Information DMSO : 125 mg/mL (231.61 mM; Need ultrasonic)
SMILES C[C@H](CCC(NCCS(=O)(O[Na])=O)=O)[C@H]1CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C.O
Molecular Weight (g/mol) 539.7
Formula Weight 539.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.