Learn More
MedChemExpress Taspoglutide
Specifications
Specifications
| Purity | 96.85% |
| CAS | 275371-94-3 |
| Content And Storage | Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| For Use With (Application) | Metabolism-sugar/lipid metabolism |
| Molecular Weight (g/mol) | 3339.71 g/mol |
| Product Type | Peptides |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : ≥ 28 mg/mL (8.38 mM) |
| Synonym | ITM077∣ R1583∣ BIM51077 |