Learn More
Lenz Laborglasintrumente Taps for Aspirator Bottles
Product Code. 10462911
Change view
Click to view available options
Plug Bore:
4 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Bore unitSize
10462911 4 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10462911 Supplier Lenz Laborglasintrumente Supplier No. 02801904
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With solid glass plug and screwthread retaining device
  • Material: borosilicate glass 3.3
Specifications
Plug Bore 4 mm
Material Borosilicate Glass (3.3)