missing translation for 'onlineSavingsMsg'
Learn More

TAPI-2, MedChemExpress

Product Code. 16452848
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16452848

Brand: Medchem Express HY100211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TAPI-2 (TNF Protease Inhibitor 2) is a broad-spectrum inhibitor of matrix metalloprotease (MMP), tumour necrosis factorα-converting enzyme (TACE) and a disintegrin and metalloproteinase (ADAM), with an IC50 of 20 μM for MMP. TAPI-2 blocks the entry of infectious SARS-CoV.

TAPI-2 (TNF Protease Inhibitor 2) is a broad-spectrum inhibitor of matrix metalloprotease (MMP), tumour necrosis factorα-converting enzyme (TACE) and a disintegrin and metalloproteinase (ADAM), with an IC50 of 20 μM for MMP. TAPI-2 blocks the entry of infectious SARS-CoV.

Specifications

Chemical Name or Material TAPI-2
Physical Form Solid
CAS 187034-31-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H37N5O5
For Use With (Application) Cancer-Kinase/protease
Synonym TNF Protease Inhibitor 2
Solubility Information H2O : 100 mg/mL (240.66 mM; Need ultrasonic) ∣DMSO : ≥ 22 mg/mL (52.94 mM)
SMILES CC(C)(C)[C@@H](C(N[C@H](C(NCCN)=O)C)=O)NC(C(CC(NO)=O)CC(C)C)=O
Molecular Weight (g/mol) 415.53
Formula Weight 415.53
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.