Learn More
Thermo Scientific™ Tango™ AGTR1-bla U2OS Cells
Product Code. 10756875
Change view
Click to view available options
Quantity:
8 x 106 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10756875 8 x 106 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10756875 Supplier Thermo Scientific™ Supplier No. K1443
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Tango™ AGTR1-bla U2OS cells contain the human Angiotensin Receptor 1 (AGTR1) (AGTR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Tango™ AGTR1-bla U2OS cells contain the human Angiotensin Receptor 1 (AGTR1) (AGTR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ AGTR1-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of Angiotensin II (Figure 1). In addition, Tango™ AGTR1-bla U2OS cells have been tested for assay performance under variable conditions.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number NM_032049.2
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• Tango™ AGTR1-UAS-bla CHO-K1 cells
• Tango™ AGTR1-UAS-bla CHO-K1 Assay Protocol
• Certificate of Analysis
The Tango™ AGTR1-UAS-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Cytoplasmic Tail Native Tail
Description Tango™ AGTR1-bla U2OS Cells, 8 x 106 Cells, ∽8,000,000 cells/vial at 8,000,000 cells/mL Quantity of Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol AGTR1
Human Protein Class G-Protein Coupled Receptor (GPCR)
Includes Tango™ AGTR1-UAS-bla CHO-K1 cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: Tango™ AGTR1-UAS-bla CHO-K1 cells, Tango™ AGTR1-UAS-bla CHO-K1 assay protocol, certificate of analysis
Population Status Single Cell Clone Origin
Product Line Tango
Quantity 8 x 106 Cells
Readout End Point
Sample Type U2OS
Shipping Condition Room Temperature
Target Entry AGTR1
Type U2OS Cell
Cell Line U2OS
System Type Tango™
Druggable Target GPCR