missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TAMRA Monoclonal specifically detects TAMRA in Non-species specific samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
| Antigen | TAMRA |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 5G5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunoprecipitation 10 ug |
| Gene Alias | BP-25606, F07-0040, TAMRA Azide, TAMRA Azide, isomer 5, tetramethylrhodamine azide |
| Host Species | Mouse |
| Immunogen | TAMRA molecule |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?