Learn More
Buerkle™ Tamper Evident Screw Cap
Product Code. 17925171 Shop All Bürkle Products
Click to view available options
Thread Size (Metric):
42 mm
Unit Size:
Each
Product Code. 17925171
Brand: Buerkle™ 14279106
Only null left
Add to basket
Add to basket
The tamper evident closure ensures that the containers can be stored or shipped unopened. Once unscrewed, the plastic edge of the tamper-evident seal breaks off when the container is opened. This makes it possible to check whether the container has already been opened or not.
The tamper evident seal ensures that the containers can be stored or shipped unopened.
Once unscrewed, the plastic edge of the tamper evident ring breaks off when the container is opened. This makes it possible to check whether the container has already been opened or not.
The closure can then be reused indefinitely as a normal lid.
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Material HDPE
Sterility Non-sterile
Thread Size (Metric) 42 mm
Neck Finish 42 mm
Disposable Limited-use
Product Type Screw Cap
Autoclavable Not Autoclavable