missing translation for 'onlineSavingsMsg'
Learn More

TagRFP Polyclonal Antibody, Invitrogen™

Product Code. 10041338
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10041338 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10041338 Supplier Invitrogen Supplier No. R10367

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody can be used to detect native RFP, RFP variants, and RFP fusion proteins in Western blot, immunoprecipitation, ELISA, and immunocytochemistry. This antibody has been shown to detect denatured and native TurboRFP, TurboFP602, TurboFP635, TagRFP, TagFP635, and mKate2. Storage and reconstitution: to prepare a 1 mg/mL stock solution, add 100 μL sterile water or 50% glycerol to the vial. If desired, aliquot the antibody upon reconstitution to produce single-use samples. Store the reconstituted product with sterile water at 4° C, avoiding freeze/thaw cycles. When stored as directed, the solution is stable for 3 months. If reconstituted with 50% glycerol, store at -20° C, avoiding freeze/thaw cycles. When stored as directed, this solution is stable for 6 months.

TagRFP Polyclonal antibody specifically detects TagRFP in Tag samples. It is validated for ELISA, Immunocytochemistry, Western Blot.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Antigen TagRFP
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.25mg/mL gelatin, 1% trehalose and no preservative; pH 7.4
Gene Alias Entacmaea; GFP-like chromoprotein; Red fluorescent protein; RFP; RFP tag
Host Species Rabbit
Immunogen Full-length recombinant denatured and non-denatured TagRFP was used as the immunogen
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, store in dark
Product Type Antibody
Form Lyophilized
Isotype IgG
Target Molecule Tag
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.