missing translation for 'onlineSavingsMsg'
Learn More

TAF5 Antibody - BSA Free, Novus Biologicals™

Product Code. 18623560 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18623560 0.02 mL 0.02mL
18647771 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18623560 Supplier Novus Biologicals Supplier No. NBP2930250.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAF5 Polyclonal antibody specifically detects TAF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF5
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:100
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias TAF(II)100, TAF2DTATA box binding protein (TBP)-associated factor 2D, TAF5 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAFII100TAFII-100, TATA box binding protein (TBP)-associated factor, RNA polymerase II, D, 100kD, transcription initiation factor TFIID 100 kD subunit, Transcription initiation factor TFIID 100 kDa subunit, transcription initiation factor TFIID subunit 5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 656-800 of human TAF5 (NP_008882.2). VLNGNCVRIFTGHKGPIHSLTFSPNGRFLATGATDGRVLLWDIGHGLMVGELKGHTDTVCSLRFSRDGEILASGSMDNTVRLWDAIKAFEDLETDDFTTATGHINLPENSQELLLGTYMTKSTPVVHLHFTRRNLVLAAGAYSPQ
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6877
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.