missing translation for 'onlineSavingsMsg'
Learn More

TAF12 Antibody [HRP], Novus Biologicals Biologicals™

Product Code. 30500452 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30500452 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30500452 Supplier Novus Biologicals Supplier No. NBP337958H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAF12 Polyclonal antibody specifically detects TAF12 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF12
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias 20kDa, TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF15, TAF2J, TAFII-20/TAFII-15, TAFII20TAFII20/TAFII15, TATA box binding protein (TBP)-associated factor, RNA polymerase II, J, 20kD, Transcription initiation factor TFIID 20/15 kDa subunits, transcription initiation factor TFIID subunit 12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-161 of human TAF12 (NP_001128690.1).,, Sequence:, MNQFGPSALINLSNFSSIKPEPASTPPQGSMANSTAVVKIPGTPGAGGRLSPENNQVLTKKKLQDLVREVDPNEQLDEDVEEMLLQIADDFIESVVTAACQLARHRKSSTLEVKDVQLHLERQWNMWIPGFGSEEIRPYKKACTTEAHKQRMALIRKTTKK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6883
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.