missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TAF12 Polyclonal antibody specifically detects TAF12 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | TAF12 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 20kDa, TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF15, TAF2J, TAFII-20/TAFII-15, TAFII20TAFII20/TAFII15, TATA box binding protein (TBP)-associated factor, RNA polymerase II, J, 20kD, Transcription initiation factor TFIID 20/15 kDa subunits, transcription initiation factor TFIID subunit 12 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-161 of human TAF12 (NP_001128690.1).,, Sequence:, MNQFGPSALINLSNFSSIKPEPASTPPQGSMANSTAVVKIPGTPGAGGRLSPENNQVLTKKKLQDLVREVDPNEQLDEDVEEMLLQIADDFIESVVTAACQLARHRKSSTLEVKDVQLHLERQWNMWIPGFGSEEIRPYKKACTTEAHKQRMALIRKTTKK |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?