Learn More
DiscoverX TAE684
Product Code. 17342509
Encompass
Click to view available options
:
5 mg
This item is not returnable. View return policy
Product Code. 17342509
Brand: DiscoverX 921185
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
TAE684 is an inhibitor of ALK. Provided as 5mg of dry powder.
TAE684 (NVP-TAE684) is a synthetic potent and selective inhibitor of the kinase ALK as well as FLT3 and TIE2. It is 100 times more selective for ALK than InsR, and is effective at inhibiting the NPM-ALK fusion protein responsible for survival and proliferation of anaplastic large-cell lymphomas (ALCLs). Provided as 5mg of dry powder.
Specifications
Quantity 5 mg