missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAAR6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5116000
Additional Details : Weight : 0.01000kg
Description
Antibody detects endogenous levels of total TAAR6.
Trace amines are biogenic amines present in very low levels in mammalian tissues. Although some trace amines have clearly defined roles as neurotransmitters in invertebrates, the extent to which they function as true neurotransmitters in vertebrates has remained speculative. Trace amines are likely to be involved in a variety of physiological functions that have yet to be fully understood. Expressed at low abundance in various brain tissues, as well as in fetal liver, but not in the cerebellum or placenta. In the brain, comparable levels of expression in basal ganglia, frontal cortex, substantia nigra, amygdala and hippocampus, highest expression in hippocampus and lowest expression in basal ganglia.Specifications
TAAR6 | |
Polyclonal | |
Unconjugated | |
TAAR6 | |
Gm228; mTaar6; RP11-295F4.3; TA4; TAAR6; TAR4; taR-4; TAR6; taR-6; trace amine associated receptor 6; trace amine receptor 4; trace amine receptor 6; trace amine-associated receptor 6; trace-amine-associated receptor 6; TRAR4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
215855, 294124, 319100 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q5QD13, Q923Y5, Q96RI8 | |
TAAR6 | |
A synthesized peptide derived from human TAAR6(Accession Q96RI8), corresponding to amino acid residues A224-Y274. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |