Learn More
TA-02, MedChemExpress
Product Code. 16482778
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16482778
Brand: Medchem Express HY100115
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress TA-02, an analog of SB 203580 (HY-10256), is a p38 MAPK inhibitor with an IC50 of 20 nM. TA-02 especially inhibits TGFBR-2. TA-02 exhibits similar cardiogenic properties as SB 203580 and SB 202190 (HY-10295).
TA-02, an analog of SB 203580 (HY-10256), is a p38 MAPK inhibitor with an IC50 of 20 nM. TA-02 especially inhibits TGFBR-2. TA-02 exhibits similar cardiogenic properties as SB 203580 and SB 202190 (HY-10295).
Specifications
Chemical Name or Material TA-02
CAS 1784751-19-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H13F2N3
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (75.00 mM; Need ultrasonic)
SMILES FC(C=C1)=CC=C1C2=C(C3=CC=NC=C3)NC(C4=CC=CC=C4F)=N2
Molecular Weight (g/mol) 333.33
Formula Weight 333.33
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease