Learn More
Prestige Medical T-Classic Carry Bag
Product Code. 17859588
Change view
Click to view available options
For Use With (Equipment):
T-Classic 9 L Models
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17859588 T-Classic 9 L Models Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17859588 Supplier Prestige Medical Supplier No. 5258A
  • ONLINE EXCLUSIVE PRICE
    35.54 €
    33.55 € / Pack of 1
    Save 1.99 €
    5% Off
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
T-Classic Carry Bag for 9 L models only.
  • T-Classic carry bag (9 L models only)
Specifications
For Use With (Equipment) T-Classic 9 L Models
Product Type Carry Bag