Learn More
CELLPATH SYSTEM II+ 2 WELL CASSETTE
Product Code. p-200113606
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2000
This item is not returnable. View return policy
Product Code. 30226256
Brand: CELLPATH EAM170203A
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
The System II+ 2 Well Cassettes incorporate two wells, which allows multiple biopsies to process simultaneously and significantly reduce workload. This method negates using tissue wrap or biopsy bags and makes it easier to handle biopsies, which also reduces the risk of potential core fragmentation.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Material Loose
Product Type Cassette