Learn More
Trajan Scientific and Medical™ Syringe fixed 10μL SGE
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Pack of 25
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Manual injection syringe
Specifications
Specifications
| Needle Gauge | 26 G |
| Diameter (Metric) Inner Needle | 0.11 mm |
| Diameter (Metric) Outer Needle | 0.47 mm |
| Quantity | 25/Pk. |
| Volume (Metric) | 10 μL |
| Needle Point Style | Bevel |